Assay Details
Gene Symbol: CYCD2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127815.4 | NP_179835.1 | 5 - 6 | 1123 | 87 | |
GenBank mRNA | BT025242.1 | - | 5 - 6 | 829 | 87 | |
X83370.1 | - | 1023 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
