Assay Details
| Gene Symbol: SVP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161056.1 | NP_001154528.1 | 6 - 7 | 1040 | 72 | |
| NM_001335800.1 | NP_001324584.1 | 7 - 8 | 619 | 72 | ||
| NM_127820.4 | NP_179840.2 | 6 - 7 | 1282 | 72 | ||
| GenBank mRNA | AF211171.1 | - | 6 - 7 | 1041 | 72 | |
| BT033098.1 | - | 6 - 7 | 512 | 72 | ||
| EU078686.1 | - | 6 - 7 | 497 | 72 | ||
| EU078687.1 | - | 6 - 7 | 512 | 72 | ||
More Information
Set Membership: |
|

SVP
Genomic Map