Assay Details
| Gene Symbol: AGL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335805.1 | NP_001324454.1 | 5 - 6 | 629 | 161 | |
| NM_127828.3 | NP_179848.1 | 5 - 6 | 629 | 161 | ||
| GenBank mRNA | AB493558.1 | - | 5 - 6 | 464 | 161 | |
More Information
Set Membership: |
|

AGL17
Genomic Map