Assay Details
| Gene Symbol: BRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127829.3 | NP_179849.2 | 1 - 2 | 423 | 69 | |
| GenBank mRNA | AF370530.1 | - | 1 - 2 | 352 | 69 | |
| AY085644.1 | - | 1 - 2 | 297 | 69 | ||
| BT000017.1 | - | 1 - 2 | 137 | 69 | ||
More Information
Set Membership: |
|

BRK1
Genomic Map