Assay Details
| Gene Symbol: PAB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127899.4 | NP_179916.1 | 6 - 7 | 1676 | 76 | |
| GenBank mRNA | AY050859.1 | - | 6 - 7 | 1536 | 76 | |
| AY079389.1 | - | 6 - 7 | 1382 | 76 | ||
More Information
Set Membership: |
|

PAB4
Genomic Map