Assay Details
| Gene Symbol: PAB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127899.4 | NP_179916.1 | 8 - 9 | 2020 | 81 | |
| GenBank mRNA | AY050859.1 | - | 8 - 9 | 1880 | 81 | |
| AY079389.1 | - | 8 - 9 | 1726 | 81 | ||
More Information
Set Membership: |
|

PAB4
Genomic Map