Assay Details
| Gene Symbol: ICK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127907.4 | NP_179924.1 | 2 - 3 | 405 | 141 | |
| GenBank mRNA | BT028986.1 | - | 2 - 3 | 236 | 141 | |
More Information
Set Membership: |
|

ICK1
Genomic Map