Assay Details
| Gene Symbol: XLG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127910.3 | NP_565553.1 | 3 - 4 | 1741 | 74 | |
| GenBank mRNA | AF060941.1 | - | 3 - 4 | 1665 | 74 | |
More Information
Set Membership: |
|

XLG1
Genomic Map