Assay Details
| Gene Symbol: eIFiso4G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127967.4 | NP_179983.1 | 5 - 6 | 773 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eIFiso4G2| Gene Symbol: eIFiso4G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127967.4 | NP_179983.1 | 5 - 6 | 773 | 138 | |
Set Membership: |
|