Assay Details
| Gene Symbol: AT2G24060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127968.3 | NP_179984.1 | 1 - 2 | 392 | 83 | |
| GenBank mRNA | AY072160.1 | - | 1 - 2 | 341 | 83 | |
| AY088131.1 | - | 1 - 2 | 355 | 83 | ||
| AY096414.1 | - | 1 - 2 | 289 | 83 | ||
More Information
Set Membership: |
|

AT2G24060
Genomic Map