Assay Details
| Gene Symbol: AT2G24060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127968.3 | NP_179984.1 | 3 - 4 | 484 | 62 | |
| GenBank mRNA | AY072160.1 | - | 3 - 4 | 433 | 62 | |
| AY096414.1 | - | 3 - 4 | 381 | 62 | ||
More Information
Set Membership: |
|

AT2G24060
Genomic Map