Assay Details
| Gene Symbol: SCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127973.5 | NP_179989.1 | 11 - 12 | 2311 | 98 | |
| NM_201796.2 | NP_973525.2 | 11 - 12 | 2311 | 98 | ||
| GenBank mRNA | AK229652.1 | - | 11 - 12 | 2217 | 98 | |
| Y08463.1 | - | 2258 | 98 | |||
More Information
Set Membership: |
|

SCA3
Genomic Map