Assay Details
| Gene Symbol: SCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127973.5 | NP_179989.1 | 16 - 17 | 2759 | 113 | |
| GenBank mRNA | AK229652.1 | - | 16 - 17 | 2665 | 113 | |
| Y08463.1 | - | 2706 | 113 | |||
More Information
Set Membership: |
|

SCA3
Genomic Map