Assay Details
| Gene Symbol: SCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127973.5 | NP_179989.1 | 17 - 18 | 2875 | 75 | |
| GenBank mRNA | AK229652.1 | - | 17 - 18 | 2781 | 75 | |
| Y08463.1 | - | 2822 | 75 | |||
More Information
Set Membership: |
|

SCA3
Genomic Map