Assay Details
| Gene Symbol: LAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124903.1 | NP_001118375.1 | 8 - 9 | 1255 | 74 | |
| NM_001202658.1 | NP_001189587.1 | 8 - 9 | 1147 | 74 | ||
| NM_127981.4 | NP_179997.1 | 7 - 8 | 1411 | 74 | ||
| GenBank mRNA | AY062105.1 | - | 7 - 8 | 1244 | 74 | |
| BT006345.1 | - | 7 - 8 | 1176 | 74 | ||
| X63444.1 | - | 1205 | 74 | |||
More Information
Set Membership: |
|

LAP1
Genomic Map