Assay Details
| Gene Symbol: ALDH11A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202660.1 | NP_001189589.1 | 3 - 4 | 569 | 84 | |
| NM_001202661.1 | NP_001189590.1 | 3 - 4 | 580 | 84 | ||
| NM_001335927.1 | NP_001318281.1 | 3 - 4 | 647 | 84 | ||
| NM_127989.4 | NP_180004.1 | 2 - 3 | 790 | 84 | ||
| GenBank mRNA | AK317484.1 | - | 3 - 4 | 408 | 84 | |
| AY037205.1 | - | 2 - 3 | 461 | 84 | ||
| AY136409.1 | - | 2 - 3 | 546 | 84 | ||
| BT004551.1 | - | 2 - 3 | 343 | 84 | ||
More Information
Set Membership: |
|

ALDH11A3
Genomic Map