Assay Details
| Gene Symbol: ALDH11A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202660.1 | NP_001189589.1 | 9 - 10 | 1602 | 91 | |
| NM_001202661.1 | NP_001189590.1 | 9 - 10 | 1613 | 91 | ||
| NM_001335927.1 | NP_001318281.1 | 9 - 10 | 1680 | 91 | ||
| NM_127989.4 | NP_180004.1 | 8 - 9 | 1823 | 91 | ||
| GenBank mRNA | AK317484.1 | - | 9 - 10 | 1441 | 91 | |
| AY037205.1 | - | 8 - 9 | 1494 | 91 | ||
| AY136409.1 | - | 8 - 9 | 1579 | 91 | ||
| BT004551.1 | - | 8 - 9 | 1376 | 91 | ||
More Information
Set Membership: |
|

ALDH11A3
Genomic Map