Assay Details
| Gene Symbol: DEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114685.4 | NP_190399.2 | 1 - 2 | 438 | 92 | |
| NM_180681.1 | NP_851012.1 | 1 - 2 | 438 | 92 | ||
| GenBank mRNA | AB074533.1 | - | 1 - 2 | 128 | 92 | |
| AJ417836.1 | - | 1 - 2 | 135 | 92 | ||
| BT004258.1 | - | 1 - 2 | 438 | 92 | ||
| BT005524.1 | - | 1 - 2 | 128 | 92 | ||
More Information
Set Membership: |
|

DEL1
Genomic Map