Assay Details
| Gene Symbol: DEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114685.4 | NP_190399.2 | 4 - 5 | 644 | 140 | |
| NM_180681.1 | NP_851012.1 | 4 - 5 | 644 | 140 | ||
| GenBank mRNA | AB074533.1 | - | 4 - 5 | 334 | 140 | |
| AJ417836.1 | - | 4 - 5 | 341 | 140 | ||
| BT004258.1 | - | 4 - 5 | 644 | 140 | ||
| BT005524.1 | - | 4 - 5 | 334 | 140 | ||
More Information
Set Membership: |
|

DEL1
Genomic Map