Assay Details
| Gene Symbol: AT3G48860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339384.1 | NP_001325715.1 | 7 - 8 | 1413 | 106 | |
| NM_114745.6 | NP_190455.2 | 8 - 9 | 1188 | 106 | ||
| NM_180683.3 | NP_851014.1 | 8 - 9 | 1045 | 106 | ||
| GenBank mRNA | AY093166.1 | - | 8 - 9 | 1045 | 106 | |
| BT033150.1 | - | 8 - 9 | 912 | 106 | ||
| KC533769.1 | - | 8 - 9 | 912 | 106 | ||
More Information
Set Membership: |
|

AT3G48860
Genomic Map