Assay Details
| Gene Symbol: DFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339736.1 | NP_001327282.1 | 9 - 10 | 958 | 66 | |
| NM_001339737.1 | NP_001327281.1 | 7 - 8 | 1057 | 66 | ||
| NM_001339738.1 | NP_001327280.1 | 8 - 9 | 991 | 66 | ||
| NM_001339739.1 | NP_001327279.1 | 9 - 10 | 720 | 66 | ||
| NM_115421.4 | NP_567026.3 | 10 - 11 | 879 | 66 | ||
| NM_180686.2 | NP_851017.1 | 10 - 11 | 754 | 66 | ||
| NM_180687.2 | NP_851018.1 | 10 - 11 | 809 | 66 | ||
| GenBank mRNA | AJ292545.1 | - | 10 - 11 | 710 | 66 | |
| AY050867.1 | - | 10 - 11 | 781 | 66 | ||
| AY091206.1 | - | 9 - 10 | 698 | 66 | ||
More Information
Set Membership: |
|

DFD
Genomic Map