Assay Details
| Gene Symbol: DFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339736.1 | NP_001327282.1 | 12 - 13 | 1397 | 86 | |
| NM_001339737.1 | NP_001327281.1 | 10 - 11 | 1496 | 86 | ||
| NM_001339738.1 | NP_001327280.1 | 11 - 12 | 1430 | 86 | ||
| NM_001339739.1 | NP_001327279.1 | 12 - 13 | 1159 | 86 | ||
| NM_115421.4 | NP_567026.3 | 13 - 14 | 1318 | 86 | ||
| NM_180686.2 | NP_851017.1 | 13 - 14 | 1193 | 86 | ||
| NM_180687.2 | NP_851018.1 | 13 - 14 | 1248 | 86 | ||
| GenBank mRNA | AJ292545.1 | - | 13 - 14 | 1149 | 86 | |
| AY050867.1 | - | 13 - 14 | 1220 | 86 | ||
| AY091206.1 | - | 12 - 13 | 1137 | 86 | ||
More Information
Set Membership: |
|

DFD
Genomic Map