Assay Details
| Gene Symbol: DFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339736.1 | NP_001327282.1 | 7 - 8 | 838 | 86 | |
| NM_001339737.1 | NP_001327281.1 | 5 - 6 | 937 | 86 | ||
| NM_001339738.1 | NP_001327280.1 | 6 - 7 | 871 | 86 | ||
| NM_001339739.1 | NP_001327279.1 | 7 - 8 | 600 | 86 | ||
| NM_115421.4 | NP_567026.3 | 8 - 9 | 759 | 86 | ||
| NM_180686.2 | NP_851017.1 | 8 - 9 | 634 | 86 | ||
| NM_180687.2 | NP_851018.1 | 8 - 9 | 689 | 86 | ||
| GenBank mRNA | AJ292545.1 | - | 8 - 9 | 590 | 86 | |
| AY050867.1 | - | 8 - 9 | 661 | 86 | ||
| AY091206.1 | - | 7 - 8 | 578 | 86 | ||
More Information
Set Membership: |
|

DFD
Genomic Map