Assay Details
| Gene Symbol: AT3G63460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203242.1 | NP_001190171.1 | 12 - 13 | 1404 | 65 | |
| NM_116211.3 | NP_191905.3 | 12 - 13 | 1398 | 65 | ||
| NM_180693.2 | NP_851024.1 | 12 - 13 | 1427 | 65 | ||
| GenBank mRNA | AY099702.1 | - | 12 - 13 | 1368 | 65 | |
| BT008435.1 | - | 12 - 13 | 1301 | 65 | ||
More Information
Set Membership: |
|

AT3G63460
Genomic Map