Assay Details
| Gene Symbol: SVR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121368.4 | NP_568289.3 | 12 - 13 | 1944 | 63 | |
| NM_180704.2 | NP_851035.1 | 12 - 13 | 1773 | 63 | ||
More Information
Set Membership: |
|

SVR3
Genomic Map