Assay Details
| Gene Symbol: SVR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121368.4 | NP_568289.3 | 3 - 4 | 770 | 62 | |
| NM_180704.2 | NP_851035.1 | 3 - 4 | 599 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SVR3| Gene Symbol: SVR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121368.4 | NP_568289.3 | 3 - 4 | 770 | 62 | |
| NM_180704.2 | NP_851035.1 | 3 - 4 | 599 | 62 | ||
Set Membership: |
|