Assay Details
| Gene Symbol: DIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036833.2 | NP_001031910.1 | 2 - 3 | 1959 | 98 | |
| NM_001203418.1 | NP_001190347.1 | 3 - 4 | 1866 | 98 | ||
| NM_122032.6 | NP_197525.1 | 3 - 4 | 1945 | 98 | ||
| NM_180713.4 | NP_851044.2 | 2 - 3 | 2044 | 98 | ||
| GenBank mRNA | AK227214.1 | - | 3 - 4 | 1849 | 98 | |
| AK316714.1 | - | 3 - 4 | 1711 | 98 | ||
| AK318923.1 | - | 2 - 3 | 1949 | 98 | ||
| AY090237.1 | - | 3 - 4 | 1852 | 98 | ||
| BT000632.1 | - | 3 - 4 | 1795 | 98 | ||
More Information
Set Membership: |
|

DIN10
Genomic Map