Assay Details
| Gene Symbol: DIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036833.2 | NP_001031910.1 | 3 - 4 | 2094 | 77 | |
| NM_001203418.1 | NP_001190347.1 | 4 - 5 | 2001 | 77 | ||
| NM_122032.6 | NP_197525.1 | 4 - 5 | 2080 | 77 | ||
| NM_180713.4 | NP_851044.2 | 3 - 4 | 2179 | 77 | ||
| GenBank mRNA | AK227214.1 | - | 4 - 5 | 1984 | 77 | |
| AK316714.1 | - | 4 - 5 | 1846 | 77 | ||
| AK318923.1 | - | 3 - 4 | 2084 | 77 | ||
| AY090237.1 | - | 4 - 5 | 1987 | 77 | ||
| BT000632.1 | - | 4 - 5 | 1930 | 77 | ||
More Information
Set Membership: |
|

DIN10
Genomic Map