Assay Details
| Gene Symbol: UBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343718.1 | NP_001318617.1 | 8 - 9 | 1279 | 121 | |
| NM_122126.5 | NP_568411.4 | 10 - 11 | 1405 | 121 | ||
More Information
Set Membership: |
|

UBP8
Genomic Map