Assay Details
| Gene Symbol: NF-YB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036850.1 | NP_001031927.1 | 2 - 3 | 277 | 83 | |
| NM_001343789.1 | NP_001330452.1 | 2 - 3 | 283 | 83 | ||
| NM_180729.3 | NP_851060.1 | 2 - 3 | 358 | 83 | ||
| NM_180730.3 | NP_851061.1 | 2 - 3 | 317 | 83 | ||
| GenBank mRNA | AK317460.1 | - | 2 - 3 | 229 | 83 | |
| AY057709.1 | - | 2 - 3 | 230 | 83 | ||
| AY116960.1 | - | 1 - 2 | 61 | 83 | ||
| D38110.1 | - | 2 - 3 | 161 | 83 | ||
More Information
Set Membership: |
|

NF-YB13
Genomic Map