Assay Details
| Gene Symbol: NF-YB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343789.1 | NP_001330452.1 | 4 - 5 | 535 | 117 | |
| NM_122215.3 | NP_197700.2 | 4 - 5 | 403 | 117 | ||
| NM_180729.3 | NP_851060.1 | 4 - 5 | 610 | 117 | ||
| NM_180730.3 | NP_851061.1 | 4 - 5 | 569 | 117 | ||
| GenBank mRNA | AK317460.1 | - | 4 - 5 | 481 | 117 | |
| AY057709.1 | - | 4 - 5 | 482 | 117 | ||
| AY116960.1 | - | 3 - 4 | 313 | 117 | ||
| D38110.1 | - | 4 - 5 | 413 | 117 | ||
More Information
Set Membership: |
|

NF-YB13
Genomic Map