Assay Details
| Gene Symbol: SHN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180742.1 | NP_851073.1 | 1 - 2 | 215 | 62 | |
| GenBank mRNA | AK118780.1 | - | 1 - 2 | 216 | 62 | |
| BT005461.1 | - | 1 - 2 | 80 | 62 | ||
More Information
Set Membership: |
|

SHN3
Genomic Map