Assay Details
| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 2 - 3 | 476 | 106 | |
| NM_180747.2 | NP_851078.1 | 2 - 3 | 430 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HDA2| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 2 - 3 | 476 | 106 | |
| NM_180747.2 | NP_851078.1 | 2 - 3 | 430 | 106 | ||
Set Membership: |
|