Assay Details
| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 8 - 9 | 930 | 117 | |
| NM_180747.2 | NP_851078.1 | 8 - 9 | 800 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HDA2| Gene Symbol: HDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122505.4 | NP_568480.2 | 8 - 9 | 930 | 117 | |
| NM_180747.2 | NP_851078.1 | 8 - 9 | 800 | 117 | ||
Set Membership: |
|