Assay Details
| Gene Symbol: SHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343982.1 | NP_001331385.1 | 13 - 14 | 1452 | 96 | |
| NM_122560.2 | NP_568488.2 | 13 - 14 | 1442 | 96 | ||
| NM_180749.4 | NP_851080.1 | 13 - 14 | 1452 | 96 | ||
| NM_180750.2 | NP_851081.1 | 13 - 14 | 1442 | 96 | ||
| GenBank mRNA | AK318672.1 | - | 13 - 14 | 1386 | 96 | |
| BT001097.1 | - | 13 - 14 | 1338 | 96 | ||
| BT001113.1 | - | 13 - 14 | 1338 | 96 | ||
More Information
Set Membership: |
|

SHM2
Genomic Map