Assay Details
| Gene Symbol: SHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122560.2 | NP_568488.2 | 11 - 12 | 1202 | 86 | |
| NM_180750.2 | NP_851081.1 | 11 - 12 | 1202 | 86 | ||
| GenBank mRNA | BT001097.1 | - | 11 - 12 | 1098 | 86 | |
| BT001113.1 | - | 11 - 12 | 1098 | 86 | ||
More Information
Set Membership: |
|

SHM2
Genomic Map