Assay Details
| Gene Symbol: MK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203482.1 | NP_001190411.1 | 4 - 5 | 702 | 67 | |
| NM_122627.6 | NP_198097.1 | 3 - 4 | 1253 | 67 | ||
| NM_180753.4 | NP_851084.1 | 4 - 5 | 720 | 67 | ||
| GenBank mRNA | AY084333.1 | - | 4 - 5 | 698 | 67 | |
| AY093165.1 | - | 4 - 5 | 677 | 67 | ||
| BT002104.1 | - | 3 - 4 | 630 | 67 | ||
| X77793.1 | - | 952 | 67 | |||
More Information
Set Membership: |
|

MK
Genomic Map