Assay Details
| Gene Symbol: MK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203482.1 | NP_001190411.1 | 5 - 6 | 1107 | 59 | |
| NM_122627.6 | NP_198097.1 | 4 - 5 | 1658 | 59 | ||
| NM_180753.4 | NP_851084.1 | 5 - 6 | 1125 | 59 | ||
| GenBank mRNA | AY084333.1 | - | 5 - 6 | 1103 | 59 | |
| AY093165.1 | - | 5 - 6 | 1082 | 59 | ||
| BT002104.1 | - | 4 - 5 | 1035 | 59 | ||
| X77793.1 | - | 1357 | 59 | |||
More Information
Set Membership: |
|

MK
Genomic Map