Assay Details
| Gene Symbol: PHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085160.2 | NP_001078629.1 | 5 - 6 | 967 | 101 | |
| NM_122784.5 | NP_568512.3 | 4 - 5 | 1519 | 101 | ||
| NM_180759.4 | NP_851090.1 | 5 - 6 | 865 | 101 | ||
| GenBank mRNA | BT002344.1 | - | 4 - 5 | 883 | 101 | |
More Information
Set Membership: |
|

PHL1
Genomic Map