Assay Details
| Gene Symbol: BR6OX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344281.1 | NP_001332353.1 | 2 - 3 | 624 | 86 | |
| NM_123259.4 | NP_198713.3 | 1 - 2 | 901 | 86 | ||
| NM_180774.2 | NP_851105.1 | 2 - 3 | 835 | 86 | ||
| NM_203133.2 | NP_974862.1 | 2 - 3 | 656 | 86 | ||
| GenBank mRNA | AB035868.1 | - | 2 - 3 | 631 | 86 | |
More Information
Set Membership: |
|

BR6OX1
Genomic Map