Assay Details
| Gene Symbol: BR6OX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344281.1 | NP_001332353.1 | 3 - 4 | 777 | 73 | |
| NM_123259.4 | NP_198713.3 | 2 - 3 | 1054 | 73 | ||
| NM_180774.2 | NP_851105.1 | 3 - 4 | 988 | 73 | ||
| NM_203133.2 | NP_974862.1 | 3 - 4 | 809 | 73 | ||
| GenBank mRNA | AB035868.1 | - | 3 - 4 | 784 | 73 | |
More Information
Set Membership: |
|

BR6OX1
Genomic Map