Assay Details
| Gene Symbol: BR6OX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344281.1 | NP_001332353.1 | 8 - 9 | 1413 | 127 | |
| NM_123259.4 | NP_198713.3 | 7 - 8 | 1690 | 127 | ||
| NM_180774.2 | NP_851105.1 | 8 - 9 | 1624 | 127 | ||
| NM_203133.2 | NP_974862.1 | 8 - 9 | 1444 | 127 | ||
| GenBank mRNA | AB035868.1 | - | 8 - 9 | 1420 | 127 | |
More Information
Set Membership: |
|

BR6OX1
Genomic Map