Assay Details
| Gene Symbol: SYN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125872.1 | NP_001119344.1 | 6 - 7 | 1342 | 61 | |
| NM_001344360.1 | NP_001330091.1 | 6 - 7 | 1399 | 61 | ||
| NM_001344361.1 | NP_001330092.1 | 5 - 6 | 1577 | 61 | ||
| NM_001344362.1 | NP_001330093.1 | 6 - 7 | 1406 | 61 | ||
| NM_123449.3 | NP_568586.2 | 6 - 7 | 1463 | 61 | ||
| NM_180779.2 | NP_851110.1 | 6 - 7 | 1463 | 61 | ||
| GenBank mRNA | AF400126.1 | - | 6 - 7 | 1377 | 61 | |
| AF400127.1 | - | 6 - 7 | 1377 | 61 | ||
More Information
Set Membership: |
|

SYN2
Genomic Map