Assay Details
| Gene Symbol: DL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203519.1 | NP_001190448.1 | 10 - 11 | 1206 | 108 | |
| NM_001344434.1 | NP_001331306.1 | 10 - 11 | 1403 | 108 | ||
| NM_123573.3 | NP_568602.3 | 10 - 11 | 1202 | 108 | ||
| NM_180789.3 | NP_851120.1 | 10 - 11 | 1465 | 108 | ||
| GenBank mRNA | BT025994.1 | - | 10 - 11 | 985 | 108 | |
| L38614.1 | - | 10 - 11 | 1191 | 108 | ||
More Information
Set Membership: |
|

DL1
Genomic Map