Assay Details
| Gene Symbol: DL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344434.1 | NP_001331306.1 | 13 - 14 | 1756 | 104 | |
| NM_180789.3 | NP_851120.1 | 13 - 14 | 1818 | 104 | ||
| GenBank mRNA | L38614.1 | - | 13 - 14 | 1544 | 104 | |
More Information
Set Membership: |
|

DL1
Genomic Map