Assay Details
| Gene Symbol: DL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203519.1 | NP_001190448.1 | 14 - 15 | 1764 | 67 | |
| NM_001344434.1 | NP_001331306.1 | 14 - 15 | 1979 | 67 | ||
| NM_180789.3 | NP_851120.1 | 14 - 15 | 2041 | 67 | ||
| GenBank mRNA | L38614.1 | - | 14 - 15 | 1767 | 67 | |
More Information
Set Membership: |
|

DL1
Genomic Map