Assay Details
| Gene Symbol: COR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036921.2 | NP_001031998.1 | 1 - 2 | 184 | 151 | |
| NM_123655.5 | NP_568615.3 | 1 - 2 | 301 | 151 | ||
| NM_180790.2 | NP_851121.1 | 1 - 2 | 184 | 151 | ||
| GenBank mRNA | AK118095.1 | - | 1 - 2 | 155 | 151 | |
| AK175580.1 | - | 1 - 2 | 155 | 151 | ||
| AY088816.1 | - | 1 - 2 | 185 | 151 | ||
| BT005599.1 | - | 1 - 2 | 103 | 151 | ||
More Information
Set Membership: |
|

COR27
Genomic Map