Assay Details
| Gene Symbol: COR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036921.2 | NP_001031998.1 | 4 - 5 | 766 | 104 | |
| NM_123655.5 | NP_568615.3 | 4 - 5 | 886 | 104 | ||
| NM_180790.2 | NP_851121.1 | 4 - 5 | 769 | 104 | ||
| GenBank mRNA | AK118095.1 | - | 4 - 5 | 740 | 104 | |
| AK175580.1 | - | 4 - 5 | 740 | 104 | ||
| AY088816.1 | - | 4 - 5 | 770 | 104 | ||
| BT005599.1 | - | 4 - 5 | 688 | 104 | ||
More Information
Set Membership: |
|

COR27
Genomic Map