Assay Details
| Gene Symbol: CRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123779.5 | NP_199225.1 | 3 - 4 | 1101 | 80 | |
| NM_180796.1 | NP_851127.1 | 2 - 3 | 1170 | 80 | ||
| NM_180797.1 | NP_851128.1 | 3 - 4 | 1038 | 80 | ||
| GenBank mRNA | AY070730.1 | - | 3 - 4 | 1033 | 80 | |
| BT029491.1 | - | 2 - 3 | 699 | 80 | ||
More Information
Set Membership: |
|

CRA1
Genomic Map