Assay Details
| Gene Symbol: LARP6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085252.1 | NP_001078721.1 | 5 - 6 | 726 | 122 | |
| NM_123994.4 | NP_568660.3 | 5 - 6 | 726 | 122 | ||
| NM_180810.3 | NP_851141.1 | 5 - 6 | 757 | 122 | ||
| GenBank mRNA | AY050403.1 | - | 5 - 6 | 719 | 122 | |
| BT000588.1 | - | 5 - 6 | 649 | 122 | ||
More Information
Set Membership: |
|

LARP6a
Genomic Map